Product Details

SNP ID
rs115773666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108959056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAATTGTTCTGGAAGAGAAGA[C/T]TATCTTCTTTAAGCAAAGCTTCTAA
Phenotype
MIM: 605128 MIM: 603205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
There are no transcripts associated with this gene.

Gene
MORC1
Gene Name
MORC family CW-type zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014429.3 2998 Missense Mutation AAT,AGT N955S NP_055244.3
XM_005247362.2 2998 Missense Mutation AAT,AGT N985S XP_005247419.1
XM_011512691.1 2998 Missense Mutation AAT,AGT N987S XP_011510993.1
XM_011512692.1 2998 Missense Mutation AAT,AGT N957S XP_011510994.1
XM_011512693.1 2998 Missense Mutation AAT,AGT N922S XP_011510995.1
XM_011512694.1 2998 Missense Mutation AAT,AGT N888S XP_011510996.1
XM_011512696.2 2998 Missense Mutation AAT,AGT N761S XP_011510998.1
XM_011512697.2 2998 Missense Mutation AAT,AGT N736S XP_011510999.1
XM_017006169.1 2998 Intron XP_016861658.1
XM_017006170.1 2998 Missense Mutation AAT,AGT N706S XP_016861659.1

View Full Product Details