Product Details

SNP ID
rs116839707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128054157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGAGTTGGTTTAGAGGCTTTTC[A/G]CATCAGAGGATGAACTGTGGTACTG
Phenotype
MIM: 609213
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC61A1 PubMed Links

Gene Details

Gene
SEC61A1
Gene Name
Sec61 translocon alpha 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013336.3 Intron NP_037468.1

View Full Product Details