Product Details

SNP ID
rs113298510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151740830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAAACCCGAAGGCGAGCTGTGA[C/T]ATATTTTCATGGTGGTGGTTTTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AADACL2 PubMed Links

Gene Details

Gene
AADACL2
Gene Name
arylacetamide deacetylase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207365.3 443 Missense Mutation ACA,ATA T108I NP_997248.2
XM_011512753.1 443 Missense Mutation ACA,ATA T61I XP_011511055.1

View Full Product Details