Product Details

SNP ID
rs116611780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186576910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCACCTGAGTTGGTTGGTGGGA[A/G]AGGGAGTTATATAGTTATCATCTTC
Phenotype
MIM: 611341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB11 PubMed Links

Gene Details

Gene
DNAJB11
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016306.5 Intron NP_057390.1
Gene
TBCCD1
Gene Name
TBCC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details