Product Details

SNP ID
rs113780894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:6716700 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCAAGGCCGAGGCCGAGCTGG[C/G]CACCTTCCCCAGGGCGTTCAAGAAG
Phenotype
MIM: 605695
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BLOC1S4 PubMed Links

Gene Details

Gene
BLOC1S4
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018366.2 586 Missense Mutation GCC,GGC A164G NP_060836.1
Gene
MRFAP1L1
Gene Name
Morf4 family associated protein 1 like 1
There are no transcripts associated with this gene.

View Full Product Details