Product Details

SNP ID
rs113970135
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48138913 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACATGGATCTTACCAAATGACC[A/G]GACATCTGATTTGCTGCTGAAGCGG
Phenotype
MIM: 600583 MIM: 600058
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEC PubMed Links

Gene Details

Gene
TEC
Gene Name
tec protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003215.2 2942 Missense Mutation CGG,TGG R549W NP_003206.2
XM_011513737.2 2942 Missense Mutation CGG,TGG R513W XP_011512039.1
XM_011513741.1 2942 Intron XP_011512043.1
XM_017008569.1 2942 Missense Mutation CGG,TGG R433W XP_016864058.1
Gene
TXK
Gene Name
TXK tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details