Product Details

SNP ID
rs115250545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48848066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTGAGCGTTTTTTTTTTTGGTG[C/T]TATAATTAATTCTGTCTTCTAAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCIAD1 PubMed Links

Gene Details

Gene
OCIAD1
Gene Name
OCIA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079839.2 Intron NP_001073308.1
NM_001079840.2 Intron NP_001073309.1
NM_001079841.2 Intron NP_001073310.1
NM_001079842.2 Intron NP_001073311.2
NM_001168254.1 Intron NP_001161726.1
NM_017830.3 Intron NP_060300.1
XM_017008329.1 Intron XP_016863818.1

View Full Product Details