Product Details

SNP ID
rs114197104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17798982 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGGTATCACTGGCGGACAGATA[C/T]GTTACCAGCTTGCACAGTTAACTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017741.3 6105 Intron NP_060211.3
XM_005248169.3 6105 UTR 3 XP_005248226.1
XM_005248171.3 6105 UTR 3 XP_005248228.1
XM_006713966.3 6105 UTR 3 XP_006714029.1
XM_017008327.1 6105 UTR 3 XP_016863816.1
XM_017008328.1 6105 UTR 3 XP_016863817.1

View Full Product Details