Product Details

SNP ID
rs117918201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10564758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCGCGCCGCCTGCGCCAACAA[C/T]GTGGGGCTGCTGCGGACGCTGGTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD33B PubMed Links

Gene Details

Gene
ANKRD33B
Gene Name
ankyrin repeat domain 33B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164440.1 436 Silent Mutation AAC,AAT N97N NP_001157912.1
XM_005248343.4 436 Intron XP_005248400.1
XM_005248344.3 436 UTR 5 XP_005248401.1

View Full Product Details