Product Details
- SNP ID
-
rs116489971
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:19474190 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAGGAGCCTTAACGTTAAACCC[G/T]TTAAATTTTGATGCTTTGTAGAAAA
- Phenotype
-
MIM: 603019
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDH18
PubMed Links
Gene Details
- Gene
- CDH18
- Gene Name
- cadherin 18
View Full Product Details