Product Details

SNP ID
rs117232505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATACCTCAGGCTGGATAAACA[A/G]GCCTGAAGGGGACATGTTGTATATT
Phenotype
MIM: 604796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 1338 UTR 3 NP_062545.1

View Full Product Details