Product Details

SNP ID
rs115815829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCCCTGCCAGGAACCCTGCTGC[C/G]GGCCACTGTGTTTCTCATTTGCCGG
Phenotype
MIM: 604796
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 959 Missense Mutation CGC,GGC R214G NP_062545.1

View Full Product Details