Product Details
- SNP ID
-
rs112913377
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:81304797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGTTAGTACCACATATAACAAG[A/G]GACCCTTGCCTGCAACATCATGGGA
- Phenotype
-
MIM: 614315
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACOT12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs16878594] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACOT12
- Gene Name
- acyl-CoA thioesterase 12
There are no transcripts associated with this gene.
- Gene
- CKMT2-AS1
- Gene Name
- CKMT2 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- ZCCHC9
- Gene Name
- zinc finger CCHC-type containing 9
View Full Product Details