Product Details

SNP ID
rs117090714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:90459268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTCTGTTAATATAATTGAACCA[A/G]TCATACCCTCCTTTTTGATTGAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBLAC2 PubMed Links

Gene Details

Gene
MBLAC2
Gene Name
metallo-beta-lactamase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203406.1 3215 UTR 3 NP_981951.1

View Full Product Details