Product Details

SNP ID
rs114601508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10982457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTGACTTCATCGAAGCTAACAA[A/G]TTTTATTTTATAGGGTATAAATTAA
Phenotype
MIM: 611814 MIM: 616799
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELOVL2 PubMed Links

Gene Details

Gene
ELOVL2
Gene Name
ELOVL fatty acid elongase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017770.3 2376 UTR 3 NP_060240.3
XM_011514716.2 2376 UTR 3 XP_011513018.1
XM_011514717.2 2376 UTR 3 XP_011513019.1
XM_017010985.1 2376 Intron XP_016866474.1
Gene
SYCP2L
Gene Name
synaptonemal complex protein 2 like
There are no transcripts associated with this gene.

View Full Product Details