Product Details

SNP ID
rs113141128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3256403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTATGGATGGTTCCCCACTCGA[A/G]AAGGACTGTGATGACACTTCAGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMG4 PubMed Links

Gene Details

Gene
PSMG4
Gene Name
proteasome assembly chaperone 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128591.1 Intron NP_001122063.1
NM_001128592.1 Intron NP_001122064.1
NM_001135750.1 Intron NP_001129222.1
XM_011514591.2 Intron XP_011512893.1
XM_011514593.2 Intron XP_011512895.1
XM_011514594.2 Intron XP_011512896.1
XM_011514595.2 Intron XP_011512897.1

View Full Product Details