Product Details

SNP ID
rs114823699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37635400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAATGGACTCTGCACAGATGGGA[C/T]AGTTAAGGAACAATACCCGACAGAC
Phenotype
MIM: 609626
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDGA1 PubMed Links

Gene Details

Gene
MDGA1
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153487.3 4977 UTR 3 NP_705691.1
XM_006715056.3 4977 Intron XP_006715119.1
XM_017010734.1 4977 UTR 3 XP_016866223.1

View Full Product Details