Product Details

SNP ID
rs114192244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:86878551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTATGGCTTGGAACATACATAT[G/T]GAACACAGTGGAAGATGAGGAGAAA
Phenotype
MIM: 614048
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1324L PubMed Links

Gene Details

Gene
KIAA1324L
Gene Name
KIAA1324 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142749.2 5195 UTR 3 NP_001136221.1
NM_001291990.1 5195 UTR 3 NP_001278919.1
NM_001291991.1 5195 UTR 3 NP_001278920.1
NM_152748.3 5195 UTR 3 NP_689961.3
XM_006715894.3 5195 UTR 3 XP_006715957.1
XM_011515918.2 5195 UTR 3 XP_011514220.1
XM_011515921.1 5195 UTR 3 XP_011514223.1
XM_017011843.1 5195 UTR 3 XP_016867332.1
XM_017011844.1 5195 UTR 3 XP_016867333.1

View Full Product Details