Product Details

SNP ID
rs114451645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87840817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGATGTCACCAACATCCATCA[G/T]ATTCATTTGTAGAGGTTTGTAACCA
Phenotype
MIM: 610821
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RUNDC3B PubMed Links

Gene Details

Gene
RUNDC3B
Gene Name
RUN domain containing 3B
There are no transcripts associated with this gene.

Gene
SLC25A40
Gene Name
solute carrier family 25 member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018843.3 Intron NP_061331.2

View Full Product Details