Product Details

SNP ID
rs118072529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:97119549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTGTTTCCCCCTGTGTTAGCA[C/T]CTTACATAATCAGGGTACAGTGATC
Phenotype
MIM: 615773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDHAF3 PubMed Links

Gene Details

Gene
SDHAF3
Gene Name
succinate dehydrogenase complex assembly factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020186.2 Intron NP_064571.1

View Full Product Details