Product Details

SNP ID
rs112590068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103103535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCTAGAGCTTCATTCAGCTTC[A/T]CTGGAGGCTCTAAGTAATGCTGGCC
Phenotype
MIM: 611864 MIM: 612334
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMC10 PubMed Links

Gene Details

Gene
ARMC10
Gene Name
armadillo repeat containing 10
There are no transcripts associated with this gene.

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 1404 Missense Mutation GAG,GTG E359V NP_001116310.1
NM_198990.4 1404 Missense Mutation GAG,GTG E359V NP_945341.3
XM_005250214.4 1404 Missense Mutation GAG,GTG E359V XP_005250271.2
XM_005250215.2 1404 Missense Mutation GAG,GTG E359V XP_005250272.1
XM_005250218.2 1404 Missense Mutation GAG,GTG E359V XP_005250275.1
XM_011515933.2 1404 Missense Mutation GAG,GTG E359V XP_011514235.1
XM_011515934.2 1404 Missense Mutation GAG,GTG E224V XP_011514236.1
XM_017011854.1 1404 Missense Mutation GAG,GTG E359V XP_016867343.1
XM_017011855.1 1404 Missense Mutation GAG,GTG E359V XP_016867344.1
XM_017011856.1 1404 Intron XP_016867345.1
XM_017011857.1 1404 Intron XP_016867346.1

View Full Product Details