Product Details

SNP ID
rs118079510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66922661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTAGTTAACTGGCACCTTCTCC[A/G]TTTAGGCTCTAGCTGCATCTGTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM248 PubMed Links

Gene Details

Gene
TMEM248
Gene Name
transmembrane protein 248
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017994.4 Intron NP_060464.1
XM_005250482.3 Intron XP_005250539.1

View Full Product Details