Product Details

SNP ID
rs114481539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107564340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTTCGTTTGCCCTCCACACTCA[C/T]AAGAGTCACGCGTTAGTTGCTACAC
Phenotype
MIM: 606821
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG5 PubMed Links

Gene Details

Gene
COG5
Gene Name
component of oligomeric golgi complex 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161520.1 175 UTR 5 NP_001154992.1
NM_006348.3 175 UTR 5 NP_006339.3
NM_181733.2 175 UTR 5 NP_859422.2
Gene
DUS4L
Gene Name
dihydrouridine synthase 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270419.1 175 Intron NP_001257348.1
NM_181581.2 175 Intron NP_853559.1
XM_005250125.4 175 Intron XP_005250182.1
XM_006715844.2 175 Intron XP_006715907.1
XM_006715845.3 175 Intron XP_006715908.1
XM_011515766.2 175 Intron XP_011514068.1
XM_011515767.2 175 Intron XP_011514069.1
XM_017011708.1 175 Intron XP_016867197.1
XM_017011709.1 175 Intron XP_016867198.1
XM_017011710.1 175 Intron XP_016867199.1
XM_017011711.1 175 Intron XP_016867200.1
XM_017011712.1 175 Intron XP_016867201.1
XM_017011713.1 175 Intron XP_016867202.1

View Full Product Details