Product Details

SNP ID
rs117156132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122319811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGAAATATGTATCTCGCTTTA[C/T]ACAGAAAACATCATTTTGATGTGAT
Phenotype
MIM: 609978 MIM: 613301
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CADPS2 PubMed Links

Gene Details

Gene
CADPS2
Gene Name
calcium dependent secretion activator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009571.3 4402 UTR 3 NP_001009571.2
NM_001167940.1 4402 UTR 3 NP_001161412.1
NM_017954.10 4402 UTR 3 NP_060424.9
XM_005250695.4 4402 UTR 3 XP_005250752.1
XM_005250696.4 4402 UTR 3 XP_005250753.1
XM_005250697.4 4402 UTR 3 XP_005250754.1
XM_005250698.4 4402 UTR 3 XP_005250755.1
XM_005250699.4 4402 UTR 3 XP_005250756.1
XM_005250700.4 4402 UTR 3 XP_005250757.1
XM_005250701.4 4402 UTR 3 XP_005250758.1
XM_005250702.4 4402 UTR 3 XP_005250759.1
XM_005250703.4 4402 UTR 3 XP_005250760.1
XM_005250704.4 4402 UTR 3 XP_005250761.1
XM_005250705.4 4402 UTR 3 XP_005250762.1
XM_005250706.4 4402 UTR 3 XP_005250763.1
XM_005250707.4 4402 UTR 3 XP_005250764.1
XM_005250708.3 4402 UTR 3 XP_005250765.1
XM_011516695.2 4402 UTR 3 XP_011514997.1
XM_011516696.2 4402 UTR 3 XP_011514998.1
XM_011516697.2 4402 UTR 3 XP_011514999.1
XM_017012794.1 4402 UTR 3 XP_016868283.1
XM_017012795.1 4402 UTR 3 XP_016868284.1
XM_017012796.1 4402 UTR 3 XP_016868285.1
XM_017012797.1 4402 UTR 3 XP_016868286.1
XM_017012798.1 4402 UTR 3 XP_016868287.1
XM_017012799.1 4402 UTR 3 XP_016868288.1
XM_017012800.1 4402 UTR 3 XP_016868289.1
Gene
FEZF1
Gene Name
FEZ family zinc finger 1
There are no transcripts associated with this gene.

Gene
FEZF1-AS1
Gene Name
FEZF1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details