Product Details

SNP ID
rs115140551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123113587 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAAAATGCATCCATGATCTGA[G/T]GCAATTCAGCAGTGATAAATTGAAT
Phenotype
MIM: 606193
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC13A1 PubMed Links

Gene Details

Gene
SLC13A1
Gene Name
solute carrier family 13 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324400.1 4245 UTR 3 NP_001311329.1
NM_022444.3 4245 UTR 3 NP_071889.2
XM_011516515.2 4245 UTR 3 XP_011514817.1
XM_011516516.2 4245 UTR 3 XP_011514818.1
XM_011516517.2 4245 UTR 3 XP_011514819.1
XM_011516518.2 4245 Intron XP_011514820.1
XM_011516519.2 4245 Intron XP_011514821.1
XM_017012554.1 4245 UTR 3 XP_016868043.1
XM_017012555.1 4245 Intron XP_016868044.1

View Full Product Details