Product Details

SNP ID
rs117593251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138467919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATCTTTTTTTTTTGTTTCAAGG[A/G]CTCCTTAGGATTTTCTATGTAGTCT
Phenotype
MIM: 603406
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM24 PubMed Links

Gene Details

Gene
TRIM24
Gene Name
tripartite motif containing 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003852.3 Intron NP_003843.3
NM_015905.2 Intron NP_056989.2

View Full Product Details