Product Details

SNP ID
rs117840373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19378522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGAATATCCTTCTCTCCTTAGAA[C/G]AAACAGTTGAAGAGATTTTAATTCA
Phenotype
MIM: 180460
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DENND4C PubMed Links

Gene Details

Gene
DENND4C
Gene Name
DENN domain containing 4C
There are no transcripts associated with this gene.

Gene
RPS6
Gene Name
ribosomal protein S6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010.2 Intron NP_001001.2

View Full Product Details