Product Details

SNP ID
rs111613071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34622721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTCTAGAGCCCAAAGGATCAA[G/T]GTACATGTCTAGACAATTTGGGGGA
Phenotype
MIM: 607387
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017363.1 Intron NP_001017363.1
XM_017014283.1 Intron XP_016869772.1
Gene
DCTN3
Gene Name
dynactin subunit 3
There are no transcripts associated with this gene.

View Full Product Details