Product Details

SNP ID
rs117532952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37515318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGGCAGTTGGGGAAGAGGAG[G/T]TGCTGGAAAAAGGAACATTTTAGAA
Phenotype
MIM: 609092
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO10 PubMed Links

Gene Details

Gene
FBXO10
Gene Name
F-box protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012166.2 Intron NP_036298.2
XM_005251439.4 Intron XP_005251496.1
XM_006716754.3 Intron XP_006716817.1
XM_017014618.1 Intron XP_016870107.1
XM_017014619.1 Intron XP_016870108.1

View Full Product Details