Product Details
- SNP ID
-
rs111582923
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:100109383 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTTATTGTTATGTCAGCTTAGA[A/C]TAAAATAATCAAGTTTTTCATAGCT
- Phenotype
-
MIM: 243305
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
INVS
PubMed Links
Gene Details
- Gene
- INVS
- Gene Name
- inversin
View Full Product Details