Product Details

SNP ID
rs111558914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110669641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCACCTCAGGGGAAAGGAAAGC[A/G]ACTTAAGAAACTGACAAACACCTGT
Phenotype
MIM: 601296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUSK PubMed Links

Gene Details

Gene
MUSK
Gene Name
muscle associated receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166280.1 Intron NP_001159752.1
NM_001166281.1 Intron NP_001159753.1
NM_005592.3 Intron NP_005583.1
XM_005251994.3 Intron XP_005252051.1
XM_005251995.3 Intron XP_005252052.1
XM_005251996.3 Intron XP_005252053.1
XM_011518708.2 Intron XP_011517010.1
XM_017014734.1 Intron XP_016870223.1

View Full Product Details