Product Details

SNP ID
rs112530036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127868514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCAGTGTGGGCTGTCCAATC[C/T]GCAGGCGGGCACCATGTCACAGGGA
Phenotype
MIM: 103000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK1 PubMed Links

Gene Details

Gene
AK1
Gene Name
adenylate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000476.2 Intron NP_000467.1
NM_001318121.1 Intron NP_001305050.1
NM_001318122.1 Intron NP_001305051.1
XM_017014427.1 Intron XP_016869916.1
XM_017014428.1 Intron XP_016869917.1
Gene
MIR4672
Gene Name
microRNA 4672
There are no transcripts associated with this gene.

View Full Product Details