Product Details

SNP ID
rs114614395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128706898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGCCGCCGTCCTTCCCACAGGG[A/C]CTGAGCTGCTGGCGGAGGAGCTACA
Phenotype
MIM: 610714
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 1009 Missense Mutation ACT,CCT T176P NP_001304855.1
NM_013355.4 1009 Missense Mutation ACT,CCT T176P NP_037487.2
XM_005251946.3 1009 Missense Mutation ACT,CCT T173P XP_005252003.1
XM_006717080.2 1009 Missense Mutation ACT,CCT T183P XP_006717143.1
XM_017014649.1 1009 Missense Mutation ACT,CCT T183P XP_016870138.1
XM_017014650.1 1009 Missense Mutation ACT,CCT T50P XP_016870139.1

View Full Product Details