Product Details

SNP ID
rs116155100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17087882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATATACCATACTCATGGGTTTA[A/G]CTACATATGGACCATTAGTGTTAGG
Phenotype
MIM: 603601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3C2A PubMed Links

Gene Details

Gene
PIK3C2A
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321378.1 7147 UTR 3 NP_001308307.1
NM_001321380.1 7147 UTR 3 NP_001308309.1
NM_002645.3 7147 UTR 3 NP_002636.2
XM_011520186.2 7147 UTR 3 XP_011518488.1
XM_017017924.1 7147 UTR 3 XP_016873413.1

View Full Product Details