Product Details

SNP ID
rs112064008
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:19225735 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTGTTTTATGTGCACTCACCTG[C/G]AATTGGGGAGCTGTAAATCCTGTGG
Phenotype
MIM: 612047
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
E2F8 PubMed Links
Additional Information
For this assay, SNP(s) [rs793274] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
E2F8
Gene Name
E2F transcription factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256371.1 2545 Missense Mutation CCA,GCA P675A NP_001243300.1
NM_001256372.1 2545 Missense Mutation CCA,GCA P675A NP_001243301.1
NM_024680.3 2545 Missense Mutation CCA,GCA P675A NP_078956.2
XM_011520367.1 2545 Missense Mutation CCA,GCA P570A XP_011518669.1

View Full Product Details