Product Details

SNP ID
rs118023591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3676345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATGATGCAGACTCAGCACCAC[A/G]CGCAGCAGGTTGGCTACACGTTTGG
Phenotype
MIM: 606372 MIM: 601021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA10 PubMed Links

Gene Details

Gene
CHRNA10
Gene Name
cholinergic receptor nicotinic alpha 10 subunit
There are no transcripts associated with this gene.

Gene
NUP98
Gene Name
nucleoporin 98
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005387.6 5416 Intron NP_005378.4
NM_016320.4 5416 Silent Mutation CGC,CGT R1739R NP_057404.2
NM_139131.4 5416 Intron NP_624357.1
NM_139132.3 5416 Silent Mutation CGC,CGT R1665R NP_624358.2

View Full Product Details