Product Details

SNP ID
rs111384663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4681834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGTTCTGATCTGTTTGGTTTTG[A/G]CACCATAGATGATGGGATTGATGAC
Phenotype
MIM: 611268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51E2 PubMed Links

Gene Details

Gene
OR51E2
Gene Name
olfactory receptor family 51 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030774.3 1122 Missense Mutation GCC,GTC A293V NP_110401.1

View Full Product Details