Product Details

SNP ID
rs112109519
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5857357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCCACCAACACAATGCTCTCCA[C/T]AGCAGTGAAGAAATGGATGAAGAAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR52E8 PubMed Links

Gene Details

Gene
OR52E8
Gene Name
olfactory receptor family 52 subfamily E member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005168.1 346 Missense Mutation ATG,GTG M116V NP_001005168.1

View Full Product Details