Product Details

SNP ID
rs112511627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:18081280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGACGTCTCTTTTCTTTGAGTT[C/T]GAAGTTTATCAGGATGTCCTTGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RERGL PubMed Links

Gene Details

Gene
RERGL
Gene Name
RERG like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286201.1 736 Missense Mutation AAA,GAA K176E NP_001273130.1
NM_024730.3 736 Missense Mutation AAA,GAA K177E NP_079006.1

View Full Product Details