Product Details

SNP ID
rs116832702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:130872877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGGTACTGGAAAAACGACCTT[C/T]GTGAAACGTCATTTGACTGGTGAAT
Phenotype
MIM: 601179
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAN PubMed Links

Gene Details

Gene
RAN
Gene Name
RAN, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300796.1 175 UTR 5 NP_001287725.1
NM_001300797.1 175 UTR 5 NP_001287726.1
NM_006325.4 175 Silent Mutation TTC,TTT F26F NP_006316.1
XM_017019772.1 175 Silent Mutation TTC,TTT F26F XP_016875261.1
XM_017019773.1 175 UTR 5 XP_016875262.1

View Full Product Details