Product Details

SNP ID
rs114793660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131718688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGTTATCTAGGAATTGACTAC[C/T]TTATGTAGTGTCATGCTGTCAGGAA
Phenotype
MIM: 601945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFSWAP PubMed Links

Gene Details

Gene
SFSWAP
Gene Name
splicing factor SWAP homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261411.1 Intron NP_001248340.1
NM_004592.3 Intron NP_004583.2
XM_011538653.1 Intron XP_011536955.1
XM_011538654.2 Intron XP_011536956.1
XM_011538655.2 Intron XP_011536957.1
XM_017019798.1 Intron XP_016875287.1
XM_017019799.1 Intron XP_016875288.1

View Full Product Details