Product Details

SNP ID
rs118101419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1947349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATTTAGCATCATTCTCATGCAA[C/T]GAAGAGAGTGATGTTATCCCTCTTT
Phenotype
MIM: 609843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCP1B PubMed Links

Gene Details

Gene
DCP1B
Gene Name
decapping mRNA 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319292.1 Intron NP_001306221.1
NM_152640.4 Intron NP_689853.3
XM_011520927.2 Intron XP_011519229.1

View Full Product Details