Product Details

SNP ID
rs118117907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:3391778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGTGATGGTACACGATAGATG[C/T]TTCAATACATGGCCATCAAATGAAT
Phenotype
MIM: 610086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRMT8 PubMed Links

Gene Details

Gene
PRMT8
Gene Name
protein arginine methyltransferase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256536.1 Intron NP_001243465.1
NM_019854.4 Intron NP_062828.3
XM_017019644.1 Intron XP_016875133.1
XM_017019645.1 Intron XP_016875134.1

View Full Product Details