Product Details

SNP ID
rs116459999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122080155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTCCCACTATGAATATTAGG[C/T]ATTTGCTGCAGGTAACTTTTTTGTC
Phenotype
MIM: 608090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MLXIP PubMed Links

Gene Details

Gene
MLXIP
Gene Name
MLX interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014938.5 Intron NP_055753.3
XM_006719290.3 Intron XP_006719353.1
XM_006719291.3 Intron XP_006719354.1
XM_006719292.3 Intron XP_006719355.1
XM_006719293.3 Intron XP_006719356.1
XM_006719294.3 Intron XP_006719357.1

View Full Product Details