Product Details

SNP ID
rs113636226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109724974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGTTGGCATCCGGAAACCACC[A/G]TGCAATCTATGGGAATGATGAACTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM222A PubMed Links

Gene Details

Gene
FAM222A
Gene Name
family with sequence similarity 222 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032829.2 Intron NP_116218.2
XM_006719654.2 Intron XP_006719717.1
XM_017020055.1 Intron XP_016875544.1
Gene
FAM222A-AS1
Gene Name
FAM222A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details