Product Details
- SNP ID
-
rs117397004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:100089144 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATGGCGGGGTTCTGGGTCGGGAC[A/G]GCACCGCTGGTCGCTGCCGGACGGC
- Phenotype
-
MIM: 232000
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105370333
PubMed Links
Gene Details
- Gene
- LOC105370333
- Gene Name
- uncharacterized LOC105370333
There are no transcripts associated with this gene.
- Gene
- PCCA
- Gene Name
- propionyl-CoA carboxylase alpha subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000282.3 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
NP_000273.2 |
NM_001127692.2 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
NP_001121164.1 |
NM_001178004.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
NP_001171475.1 |
XM_005254059.2 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_005254116.1 |
XM_011521093.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_011519395.1 |
XM_017020605.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876094.1 |
XM_017020606.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876095.1 |
XM_017020607.1 |
125 |
Intron |
|
|
XP_016876096.1 |
XM_017020608.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876097.1 |
XM_017020609.1 |
125 |
Intron |
|
|
XP_016876098.1 |
XM_017020610.1 |
125 |
Intron |
|
|
XP_016876099.1 |
XM_017020611.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876100.1 |
XM_017020612.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876101.1 |
XM_017020613.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876102.1 |
XM_017020614.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876103.1 |
XM_017020615.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876104.1 |
XM_017020616.1 |
125 |
Silent Mutation |
ACA,ACG |
T8T |
XP_016876105.1 |
XM_017020617.1 |
125 |
Intron |
|
|
XP_016876106.1 |
XM_017020618.1 |
125 |
Intron |
|
|
XP_016876107.1 |
View Full Product Details