Product Details

SNP ID
rs115397441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1348261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGGGCGGGGGTGCAAGGGCTGG[G/T]CAGCCTGTCACCCTGCACCTGTGCC
Phenotype
MIM: 604009 MIM: 607838
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199096.1 3420 Silent Mutation GGG,GGT G1069G NP_001186025.1
NM_001199097.1 3420 Silent Mutation GGG,GGT G1105G NP_001186026.1
NM_001199098.1 3420 Silent Mutation GGG,GGT G1082G NP_001186027.1
NM_001199099.1 3420 Silent Mutation GGG,GGT G1077G NP_001186028.1
NM_001286464.1 3420 Silent Mutation GGG,GGT G1122G NP_001273393.1
NM_003933.4 3420 Silent Mutation GGG,GGT G1140G NP_003924.2
XM_011522728.1 3420 Silent Mutation GGG,GGT G1157G XP_011521030.1
XM_011522729.1 3420 Intron XP_011521031.1
XM_011522730.2 3420 Intron XP_011521032.1
Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
There are no transcripts associated with this gene.

Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001410.2 3420 Intron NP_001001410.1

View Full Product Details