Product Details

SNP ID
rs113626096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1434423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCACGTTTATTTCTGGATAAA[C/T]AGTGAGGGTGTGAGCTGCTGCACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143980.1 2156 Silent Mutation CTA,CTG L663L NP_001137452.1
Gene
LOC105371045
Gene Name
uncharacterized LOC105371045
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011522780.2 2156 UTR 3 XP_011521082.1

View Full Product Details