Product Details

SNP ID
rs112031979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1992713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGCACGGCGCCAGGGCCGGCC[A/G]CGACGCAGGCGGGGGACGCGGCGCG
Phenotype
MIM: 600924 MIM: 611256 MIM: 603927
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
There are no transcripts associated with this gene.

Gene
SYNGR3
Gene Name
synaptogyrin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004209.5 573 Missense Mutation ACG,GCG T139A NP_004200.2
Gene
ZNF598
Gene Name
zinc finger protein 598
There are no transcripts associated with this gene.

View Full Product Details