Product Details

SNP ID
rs114226509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27403203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGCTGCATCTTTCTCATGAAGCA[C/T]GGGGAACGGGTCGGATGGTAAGAGG
Phenotype
MIM: 605383
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL21R PubMed Links

Gene Details

Gene
IL21R
Gene Name
interleukin 21 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021798.3 617 Intron NP_068570.1
NM_181078.2 617 Intron NP_851564.1
NM_181079.4 617 Silent Mutation CAC,CAT H11H NP_851565.4
XM_011545857.2 617 Silent Mutation CAC,CAT H11H XP_011544159.1
XM_011545858.2 617 UTR 5 XP_011544160.1
XM_017023257.1 617 Intron XP_016878746.1

View Full Product Details